Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303486.1 | NP_001290415.1 | 3 | 964 | ||
NM_001303487.1 | NP_001290416.1 | 2 | 1327 | |||
NM_001324381.1 | NP_001311310.1 | 3 | 968 | |||
NM_003631.3 | NP_003622.2 | 3 | 1394 | |||
NR_130168.1 | 3 | 1394 | ||||
NR_136752.1 | 2 | 1327 | ||||
NR_136754.1 | 2 | 1327 | ||||
NR_136755.1 | 3 | 1394 | ||||
NR_136756.1 | 3 | 1394 | ||||
XM_011540305.1 | XP_011538607.1 | 3 | 1360 | |||
XM_011540308.2 | XP_011538610.1 | 3 | 1360 | |||
GenBank mRNA | AF005043.1 | - | 4 | 1258 | ||
AK293735.1 | - | 2 | 854 | |||
AK295786.1 | - | 2 | 1286 | |||
AK302560.1 | - | 3 | 935 | |||
AY258587.1 | - | 3 | 1353 | |||
AY575848.1 | - | 3 | 948 | |||
AY575849.1 | - | 2 | 859 | |||
BC050560.1 | - | 3 | 1320 | 6064831 | ||
BC052966.1 | - | 3 | 1308 | 6042331 | ||
DQ867088.1 | - | 3 | 1172 |