Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2BK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312653.1 | NP_001299582.1 | 1 | 97 | ||
NM_080593.2 | NP_542160.1 | 1 | 97 | |||
GenBank mRNA | AJ223352.1 | - | 1 | 53 | ||
AK292062.1 | - | 1 | 94 | |||
BC000893.1 | - | 1 | 79 | 3463930 | ||
BC051872.1 | - | 1 | 77 | 6526835 | ||
BC064959.1 | - | 1 | 77 | 6093977 | ||
BC108737.1 | - | 1 | 77 | 4707078 | ||
DA873786.1 | - | 1 | 97 |
Gene Symbol: H2BFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC126339.1 | - | 1 | 87 | ||
BC126369.1 | - | 1 | 87 |
Gene Symbol: LOC102724334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723918.3 | XP_006723981.1 | 1 | 61 |