Search
Search

TNFSF10| Gene Symbol: TNFSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190942.1 | NP_001177871.1 | 3 | 449 | ||
| NM_003810.3 | NP_003801.1 | 5 | 597 | |||
| NR_033994.1 | 4 | 554 | ||||
| GenBank mRNA | AK296085.1 | - | 3 | 417 | ||
| AK312742.1 | - | 5 | 565 | |||
| BC009795.1 | - | 5 | 494 | |||
| BC032722.1 | - | 5 | 549 | 5504670 | ||
| BT019564.1 | - | 5 | 474 | |||
| CR456895.1 | - | 5 | 474 | |||
| DA041872.1 | - | 4 | 514 | |||
| EU183231.1 | - | 3 | 326 | |||
| U37518.1 | - | 5 | 561 | |||
| U57059.1 | - | 5 | 564 | |||