Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003811.3 | NP_003802.1 | 3 | 521 | ||
GenBank mRNA | AK313999.1 | - | 3 | 494 | ||
BC104805.1 | - | 3 | 511 | 8143808 | ||
BC104807.1 | - | 3 | 511 | 8143810 | ||
BM790119.1 | - | 3 | 494 | |||
U03398.1 | - | 3 | 486 |