Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303468.1 | NP_001290397.1 | 7 | 687 | ||
NM_003930.4 | NP_003921.2 | 7 | 795 | |||
XM_017012771.1 | XP_016868260.1 | 7 | 757 | |||
GenBank mRNA | AB014486.1 | - | 7 | 514 | ||
AF051323.1 | - | 7 | 653 | |||
AF072166.1 | - | 7 | 481 | |||
AJ004886.1 | - | 7 | 481 | |||
AK222885.1 | - | 7 | 641 | |||
AK299216.1 | - | 8 | 634 | |||
AK299317.1 | - | 7 | 584 | |||
AK311454.1 | - | 7 | 642 | |||
AK313209.1 | - | 7 | 641 | |||
BC002893.2 | - | 7 | 639 | 3940826 | ||
BC036044.1 | - | 7 | 642 | 5272981 |