Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREB3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052854.3 | NP_443086.1 | 10, 11 | 1743 | ||
GenBank mRNA | AB063321.1 | - | 10, 11 | 1364 | ||
AF055009.1 | - | 2, 3 | 863 | |||
AK075538.1 | - | 5, 6 | 514 | |||
AK314072.1 | - | 10, 11 | 1721 | |||
BC014097.1 | - | 10, 11 | 1386 | 4546950 | ||
BC015781.1 | - | 10, 11 | 1372 | 4856946 |