Search
Search

FDXACB1| Gene Symbol: FDXACB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138378.2 | NP_612387.1 | 5 | 1919 | ||
| NR_038364.1 | 4 | 1762 | ||||
| GenBank mRNA | AK300795.1 | - | 4 | 1393 | ||
| BC006136.1 | - | 5 | 1598 | |||
| BC127680.1 | - | 5 | 1642 | 40127680 | ||