Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AURKB SIRNA Assays ›
Gene Symbol:
AURKB
Gene Name:
aurora kinase B
Gene Aliases:
AIK2, AIM-1, AIM1, ARK2, AurB, IPL1, PPP1R48, STK12, STK5, aurkb-sv1, aurkb-sv2
Chromosome Location:
Chr.17: 8204731 - 8210768 on Build GRCh38
UniGene ID:
Hs.442658
Species:
Human
siRNA ID
HSS190048
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AURKB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256834.2
NP_001243763.1
7, 8
907
NM_001284526.1
NP_001271455.1
8, 9
983
NM_001313950.1
NP_001300879.1
8, 9
956
NM_001313951.1
NP_001300880.1
8, 9
985
NM_001313952.1
NP_001300881.1
7, 8
971
NM_001313953.1
NP_001300882.1
7, 8
763
NM_001313954.1
NP_001300883.1
8, 9
990
NM_001313955.1
NP_001300884.1
6, 7
626
NM_004217.3
NP_004208.2
8, 9
980
NR_132730.1
8, 9
838
NR_132731.1
6, 7
723
XM_011524070.2
XP_011522372.1
8, 9
833
XM_011524072.2
XP_011522374.1
7, 8
1096
XM_017025307.1
XP_016880796.1
7, 8
1023
XM_017025308.1
XP_016880797.1
7, 8
953
XM_017025309.1
XP_016880798.1
7, 8
861
XM_017025310.1
XP_016880799.1
8, 9
843
XM_017025311.1
XP_016880800.1
4, 5
520
GenBank mRNA
AB011446.1
-
8, 9
921
AB011450.1
-
8, 9
915
AB519677.1
-
7, 8
858
AB519678.1
-
7, 8
762
AB519679.1
-
6, 7
722
AF004022.1
-
8, 9
925
AF008552.1
-
7, 8
858
AF015254.1
-
8, 9
925
AK297976.1
-
7, 8
862
AK314889.1
-
8, 9
929
AY677083.1
-
6, 7
723
BC000442.2
-
8, 9
919
2820719
BC009751.2
-
8, 9
899
3677349
BC013300.2
-
8, 9
939
BC080581.1
-
8, 9
911
2819846
BT019534.1
-
7, 8
858
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
protein phosphorylation
spindle organization
sister chromatid cohesion
aging
cell proliferation
attachment of spindle microtubules to kinetochore
abscission
histone modification
protein sumoylation
anaphase-promoting complex-dependent catabolic process
spindle checkpoint
negative regulation of protein binding
positive regulation of telomere maintenance via telomerase
negative regulation of cytokinesis
positive regulation of cytokinesis
protein localization to kinetochore
cellular response to UV
cleavage furrow formation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
histone H3-S28 phosphorylation
protein autophosphorylation
mitotic spindle midzone assembly
positive regulation of telomerase activity
regulation of chromosome segregation
regulation of signal transduction by p53 class mediator
positive regulation of telomere capping
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein binding
ATP binding
histone serine kinase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-hf9rh:80/10.239.161.5:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0