Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SCARB1 SIRNA Assays ›
          Gene Symbol:
          SCARB1
          Gene Name:
          scavenger receptor class B member 1
          Gene Aliases:
          CD36L1, CLA-1, CLA1, HDLQTL6, SR-BI, SRB1
          Chromosome Location:
          Chr.12: 124777628 - 124863973 on Build GRCh38
          UniGene ID:
          Hs.731377
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS190377
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SCARB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001082959.1 NP_001076428.1 7 1248
          NM_005505.4 NP_005496.4 7 1248
          GenBank mRNA AB209436.1 - 7 1079
          AF515445.1 - 8 1310
          AK124112.1 - 7 1297
          AK302779.1 - 7 940
          AK304730.1 - 7 1138
          BC080647.1 - 7 1122 6576008
          BC093732.1 - 7 1058 7939577
          BC112037.1 - 7 1070 8327505
          BC143319.1 - 7 1058 9051824
          KU177964.1 - 7 995
          KU177965.1 - 6 879
          Z22555.1 - 7 1064

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endothelial cell proliferation
          androgen biosynthetic process
          cholesterol catabolic process
          receptor-mediated endocytosis
          positive regulation of endothelial cell migration
          positive regulation of triglyceride biosynthetic process
          positive regulation of cholesterol storage
          regulation of phosphatidylcholine catabolic process
          phospholipid transport
          lipopolysaccharide transport
          lipopolysaccharide-mediated signaling pathway
          detection of lipopolysaccharide
          cholesterol efflux
          high-density lipoprotein particle remodeling
          low-density lipoprotein particle clearance
          high-density lipoprotein particle clearance
          vitamin transmembrane transport
          wound healing
          lipoprotein metabolic process
          cholesterol homeostasis
          blood vessel endothelial cell migration
          recognition of apoptotic cell
          reverse cholesterol transport
          adhesion of symbiont to host
          viral entry into host cell
          regulation of phagocytosis
          intestinal absorption
          positive regulation of nitric-oxide synthase activity
          triglyceride homeostasis
          cholesterol import
          lipopolysaccharide binding
          virus receptor activity
          phosphatidylserine binding
          lipopolysaccharide receptor activity
          transporter activity
          protein binding
          1-phosphatidylinositol binding
          high-density lipoprotein particle binding
          low-density lipoprotein particle binding
          apolipoprotein binding
          apolipoprotein A-I binding
          protein homodimerization activity
          high-density lipoprotein particle receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0