Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ENTPD6 SIRNA Assays ›
Gene Symbol:
ENTPD6
Gene Name:
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Gene Aliases:
CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3
Chromosome Location:
Chr.20: 25195670 - 25228075 on Build GRCh38
UniGene ID:
Hs.500375
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS190422
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ENTPD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114089.3
NP_001107561.1
13
1452
NM_001247.4
NP_001238.2
14
1521
NM_001317941.2
NP_001304870.1
14
1518
NM_001322378.1
NP_001309307.1
14
1665
NM_001322379.1
NP_001309308.1
11
1028
NM_001322380.1
NP_001309309.1
12
1173
NM_001322381.1
NP_001309310.1
11
1029
NM_001322382.1
NP_001309311.1
11
986
NM_001322383.1
NP_001309312.1
15
1709
NM_001322384.1
NP_001309313.1
12
1130
NM_001322385.1
NP_001309314.1
12
1308
NM_001322386.1
NP_001309315.1
12
1350
NM_001322387.1
NP_001309316.1
13
1377
NM_001322388.1
NP_001309317.1
13
1462
NM_001322389.1
NP_001309318.1
12
1494
NM_001322390.1
NP_001309319.1
14
1541
NM_001322391.1
NP_001309320.1
13
1419
NM_001322392.1
NP_001309321.1
13
1492
NM_001322393.1
NP_001309322.1
13
1495
NM_001322394.1
NP_001309323.1
13
1449
NM_001322395.1
NP_001309324.1
13
1639
NM_001322396.1
NP_001309325.1
14
1561
NM_001322397.1
NP_001309326.1
14
1564
NM_001322398.1
NP_001309327.1
13
1596
XM_006723665.2
XP_006723728.1
15
1557
XM_011529396.1
XP_011527698.1
15
1561
XM_011529397.1
XP_011527699.1
15
1490
XM_017028129.1
XP_016883618.1
12
1271
XM_017028130.1
XP_016883619.1
12
1186
XM_017028132.1
XP_016883621.1
13
1353
XM_017028133.1
XP_016883622.1
12
1241
XM_017028134.1
XP_016883623.1
11
1038
GenBank mRNA
AF039916.1
-
14
1455
AK074691.1
-
13
1301
AK094030.1
-
3
1309
AK095708.1
-
1
639
AK124625.1
-
3
3125
AK128412.1
-
8
1886
AK293256.1
-
13
1436
AK295242.1
-
8
600
AK297954.1
-
14
1404
AK300028.1
-
11
979
AK300514.1
-
11
912
AK303786.1
-
13
1407
AL834158.1
-
13
1367
AY327581.1
-
14
1418
BC025980.2
-
14
1425
3636163
CR936750.1
-
12
1074
CR936765.1
-
14
1478
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
nucleotide phosphatase
phosphatase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
response to magnesium ion
response to calcium ion
guanosine-5'-triphosphate,3'-diphosphate diphosphatase activity
nucleoside-triphosphatase activity
uridine-diphosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0