Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4DIP SIRNA Assays ›
Gene Symbol:
PDE4DIP
LOC100996724
Gene Name:
phosphodiesterase 4D interacting protein
phosphodiesterase 4D interacting protein-like
Gene Aliases:
CMYA2, MMGL
-
Chromosome Location:
Chr.1: 148808167 - 149051277 on Build GRCh38
Chromosome Location:
Chr.1: 120471449 - 120701470 on Build GRCh38
UniGene ID:
Hs.584841
Hs.731111
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS190505
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4DIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198832.2
NP_001185761.2
26
3706
NM_001198834.3
NP_001185763.3
23
3424
NM_014644.5
NP_055459.5
23
3424
XM_005272981.3
XP_005273038.2
19
4051
XM_005277442.4
XP_005277499.2
19
4051
XM_005277443.3
XP_005277500.1
19
4051
XM_006711642.2
XP_006711705.1
19
4051
XM_006711643.2
XP_006711706.1
19
4051
XM_006711644.2
XP_006711707.1
19
4051
XM_006711646.1
XP_006711709.1
26
3713
XM_006711648.2
XP_006711711.1
19
4051
XM_006711650.3
XP_006711713.1
24
3198
XM_006711651.1
XP_006711714.1
23
3316
XM_006711652.1
XP_006711715.1
23
3316
XM_011510172.1
XP_011508474.1
26
3871
XM_011510173.1
XP_011508475.1
26
3871
XM_011510174.1
XP_011508476.1
19
4051
XM_011510175.1
XP_011508477.1
26
3871
XM_011510176.1
XP_011508478.1
26
3871
XM_011510179.1
XP_011508481.1
23
3316
XM_017002875.1
XP_016858364.1
19
4051
XM_017002876.1
XP_016858365.1
19
4051
XM_017002877.1
XP_016858366.1
19
4051
XM_017002878.1
XP_016858367.1
26
3871
XM_017002879.1
XP_016858368.1
19
4051
XM_017002880.1
XP_016858369.1
26
3871
XM_017002881.1
XP_016858370.1
19
4051
XM_017002882.1
XP_016858371.1
19
4051
XM_017002883.1
XP_016858372.1
19
4051
XM_017002884.1
XP_016858373.1
19
4051
XM_017002885.1
XP_016858374.1
19
4051
XM_017002886.1
XP_016858375.1
19
4051
XM_017002887.1
XP_016858376.1
27
4148
XM_017002888.1
XP_016858377.1
27
4121
XM_017002889.1
XP_016858378.1
27
3793
XM_017002890.1
XP_016858379.1
19
4051
XM_017002891.1
XP_016858380.1
25
3952
XM_017002892.1
XP_016858381.1
25
3583
XM_017002893.1
XP_016858382.1
26
4071
XM_017002894.1
XP_016858383.1
26
3716
XM_017002895.1
XP_016858384.1
23
3034
XM_017002896.1
XP_016858385.1
19
4051
XM_017002897.1
XP_016858386.1
19
4051
XM_017002898.1
XP_016858387.1
26
3713
XM_017002899.1
XP_016858388.1
26
3713
XM_017002900.1
XP_016858389.1
19
4051
XM_017002901.1
XP_016858390.1
19
4051
XM_017002902.1
XP_016858391.1
26
3713
XM_017002903.1
XP_016858392.1
26
3713
GenBank mRNA
AB007923.2
-
27
3585
AL833273.1
-
23
3312
BX647660.1
-
23
3352
Gene Symbol: LOC100996724
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017003064.1
XP_016858553.1
24
3618
GenBank mRNA
CR749273.1
-
4
556
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein complex assembly
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline