Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPGAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320808.1 | NP_001307737.1 | 3 | 1152 | ||
NM_014873.2 | NP_055688.1 | 3 | 563 | |||
XM_005273362.3 | XP_005273419.1 | 4 | 711 | |||
XM_011510228.2 | XP_011508530.1 | 4 | 600 | |||
XM_011510229.2 | XP_011508531.1 | 3 | 483 | |||
XM_017003004.1 | XP_016858493.1 | 2 | 1715 | |||
GenBank mRNA | AY561706.1 | - | 2 | 336 | ||
BC034621.1 | - | 3 | 1153 | 4839400 | ||
BP369253.1 | - | 3 | 447 | |||
D86960.1 | - | 3 | 563 |