Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC23A2 SIRNA Assays ›
          Gene Symbol:
          SLC23A2
          Gene Name:
          solute carrier family 23 member 2
          Gene Aliases:
          NBTL1, SLC23A1, SVCT2, YSPL2, hSVCT2
          Chromosome Location:
          Chr.20: 4852356 - 5010293 on Build GRCh38
          UniGene ID:
          Hs.516866
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS190748
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC23A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005116.5 NP_005107.4 17 2249
          NM_203327.1 NP_976072.1 17 2267
          XM_011529414.2 XP_011527716.1 17 2254
          XM_011529415.2 XP_011527717.1 16 2075
          XM_011529416.2 XP_011527718.1 16 2384
          XM_011529417.2 XP_011527719.1 16 2131
          XM_017028171.1 XP_016883660.1 17 2288
          XM_017028172.1 XP_016883661.1 17 2292
          XM_017028173.1 XP_016883662.1 17 2272
          XM_017028174.1 XP_016883663.1 17 2167
          XM_017028175.1 XP_016883664.1 13 1603
          XM_017028176.1 XP_016883665.1 14 1921
          XM_017028177.1 XP_016883666.1 14 1908
          XM_017028178.1 XP_016883667.1 16 2174
          XM_017028179.1 XP_016883668.1 16 2161
          XM_017028180.1 XP_016883669.1 12 1485
          GenBank mRNA AF058319.1 - 15 1874
          AF092511.1 - 16 2170
          AF164142.1 - 17 2267
          AJ269478.1 - 15 1874
          AJ292318.1 - 16 2105
          AK296304.1 - 14 1915
          AY380556.1 - 15 1874
          D87075.2 - 17 2249
          EF032501.1 - 15 1874

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          response to oxidative stress
          nucleobase transport
          L-ascorbic acid transport
          molecular hydrogen transport
          L-ascorbic acid metabolic process
          vitamin transmembrane transport
          sodium ion transmembrane transport
          transepithelial L-ascorbic acid transport
          L-ascorbate:sodium symporter activity
          sodium-dependent multivitamin transmembrane transporter activity
          nucleobase transmembrane transporter activity
          L-ascorbic acid transporter activity
          sodium-dependent L-ascorbate transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0