Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NR1I3 SIRNA Assays ›
Gene Symbol:
NR1I3
Gene Name:
nuclear receptor subfamily 1 group I member 3
Gene Aliases:
CAR, CAR1, MB67
Chromosome Location:
Chr.1: 161229666 - 161238623 on Build GRCh38
UniGene ID:
Hs.349642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS190753
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NR1I3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077469.2
NP_001070937.1
5, 6
752
NM_001077470.2
NP_001070938.1
4, 5
612
NM_001077471.2
NP_001070939.1
5, 6
752
NM_001077472.2
NP_001070940.1
4, 5
612
NM_001077473.2
NP_001070941.1
4, 5
612
NM_001077474.2
NP_001070942.1
5, 6
752
NM_001077475.2
NP_001070943.1
4, 5
612
NM_001077476.2
NP_001070944.1
4, 5
612
NM_001077477.2
NP_001070945.1
4, 5
612
NM_001077478.2
NP_001070946.1
5, 6
752
NM_001077479.2
NP_001070947.1
4, 5
612
NM_001077480.2
NP_001070948.1
5, 6
752
NM_001077481.2
NP_001070949.1
5, 6
752
NM_001077482.2
NP_001070950.1
5, 6
752
NM_005122.4
NP_005113.1
5, 6
752
XM_005245693.4
XP_005245750.1
4, 5
1025
XM_005245694.4
XP_005245751.1
4, 5
1024
XM_005245697.4
XP_005245754.1
5, 6
635
XM_011510237.2
XP_011508539.1
4, 5
1022
GenBank mRNA
AB307702.1
-
4, 5
549
AK316208.1
-
5, 6
744
AY572806.1
-
5, 6
643
AY572807.1
-
5, 6
572
AY572808.1
-
4, 5
503
AY572809.1
-
3, 4
333
AY572810.1
-
5, 6
643
AY572811.1
-
4, 5
503
AY572812.1
-
4, 5
503
AY572813.1
-
5, 6
643
AY572814.1
-
3, 4
333
AY572815.1
-
3, 4
333
AY572816.1
-
4, 5
503
AY572817.1
-
4, 5
503
AY572818.1
-
4, 5
503
AY572819.1
-
5, 6
643
AY572820.1
-
4, 5
413
AY572826.1
-
4, 5
473
AY572827.1
-
4, 5
503
DQ022681.1
-
4, 5
549
HQ692839.1
-
4, 5
549
HQ692841.1
-
4, 5
549
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
transcription initiation from RNA polymerase II promoter
signal transduction
androgen receptor signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
androgen receptor activity
thyroid hormone receptor activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0