Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033202.3 | NP_001028374.1 | 9 | 907 | ||
GenBank mRNA | AK078164.1 | - | 5 | 259 | ||
AK144578.1 | - | 9 | 908 | |||
AK162269.1 | - | 2 | 77 | |||
BC038606.1 | - | 9 | 929 |