Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028990.4 | NP_083266.1 | 5 | 2330 | ||
GenBank mRNA | AK018471.1 | - | 4 | 1060 | ||
AK034589.1 | - | 5 | 2299 | |||
AK051532.1 | - | 5 | 2331 | |||
AK054353.1 | - | 5 | 2287 | |||
AK152957.1 | - | 5 | 2293 | |||
AK170302.1 | - | 5 | 2307 | |||
BC007160.1 | - | 5 | 2274 | 3488466 | ||
BC024911.1 | - | 5 | 2249 | 3594306 |