Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199446.1 | NP_955517.1 | 23 | 2270 | ||
GenBank mRNA | AK078940.1 | - | 13 | 1124 | ||
BC051503.1 | - | 5 | 412 | |||
BC053105.1 | - | 23 | 2270 | 6409425 |