Search
Search

Lars2| Gene Symbol: Lars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153168.2 | NP_694808.1 | 11, 12 | 1411 | ||
| GenBank mRNA | AJ428066.1 | - | 11, 12 | 1411 | ||
| AK037012.1 | - | 11, 12 | 1418 | |||
| AK045640.1 | - | 11, 12 | 1370 | |||
| AK053667.1 | - | 11, 12 | 1406 | |||
| BC053916.1 | - | 11, 12 | 1397 | |||
| BC080303.1 | - | 11, 12 | 1298 | 5695617 | ||
| BC111892.1 | - | 11, 12 | 1382 | 40060998 | ||
| BC112412.1 | - | 11, 12 | 1380 | 40060997 | ||
| BC138206.1 | - | 11, 12 | 1385 | 8861225 | ||
| BC145982.1 | - | 11, 12 | 1385 | 40131285 | ||