Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175325.3 | NP_780534.1 | 5, 6 | 408 | ||
GenBank mRNA | AK039560.1 | - | 5, 6 | 357 | ||
AK089970.1 | - | 5, 6 | 363 | |||
AK134734.1 | - | 5, 6 | 361 | |||
BC055797.1 | - | 5, 6 | 328 | |||
BC089507.1 | - | 5, 6 | 342 | |||
BC092531.1 | - | 5, 6 | 352 | |||
BC145771.1 | - | 5, 6 | 370 | 40131074 | ||
BC145773.1 | - | 5, 6 | 370 | 40131076 |