Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142809.1 | NP_001136281.1 | 13 | 2463 | ||
NM_001142810.1 | NP_001136282.1 | 13 | 2454 | |||
NM_133987.2 | NP_598748.1 | 13 | 2478 | |||
GenBank mRNA | AB077327.1 | - | 13 | 1811 | ||
AF459435.1 | - | 13 | 2461 | |||
BC024444.1 | - | 12 | 1476 | |||
BC029000.1 | - | 11 | 1226 | |||
BC049801.1 | - | 13 | 1629 | |||
BC141067.1 | - | 14 | 2204 | 9055679 | ||
BC145319.1 | - | 14 | 1868 | 9053874 | ||
EU523148.1 | - | 9 | 1512 | |||
KC994641.1 | - | 8 | 1374 |