Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207214.3 | NP_997097.1 | 18 | 2511 | ||
GenBank mRNA | AK134562.1 | - | 18 | 2024 | ||
AK150297.1 | - | 7 | 778 | |||
AK152307.1 | - | 18 | 2240 | |||
AK157473.1 | - | 18 | 2206 | |||
AK164067.1 | - | 18 | 2233 | |||
AK167145.1 | - | 18 | 2199 | |||
AK170148.1 | - | 18 | 2205 | |||
AK171012.1 | - | 18 | 2229 | |||
BC049967.1 | - | 18 | 2168 | 3672313 |