Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181404.5 | NP_852069.4 | 5 | 3040 | ||
GenBank mRNA | AK122214.1 | - | 1 | 1406 | ||
AK147945.1 | - | 2 | 2628 | |||
BC058673.1 | - | 2 | 824 | |||
BC079563.1 | - | 1 | 2367 | 30535512 |