Search
Search

Unc5b| Gene Symbol: Unc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029770.2 | NP_084046.2 | 9 | 1684 | ||
| GenBank mRNA | AJ487853.1 | - | 9 | 1587 | ||
| AK018177.1 | - | 9 | 1684 | |||
| AK147603.1 | - | 8 | 1546 | |||
| AK154271.1 | - | 9 | 1684 | |||
| AK154287.1 | - | 9 | 1687 | |||
| AK158284.1 | - | 9 | 1684 | |||
| AK170513.1 | - | 9 | 1683 | |||
| BC048162.1 | - | 8 | 1458 | |||
| BC057560.1 | - | 8 | 1458 | 6417563 | ||