Search
Search

Pi4kb| Gene Symbol: Pi4kb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293715.1 | NP_001280644.1 | 5 | 1724 | ||
| NM_001293716.1 | NP_001280645.1 | 3 | 742 | |||
| NM_175356.3 | NP_780565.2 | 4 | 1679 | |||
| GenBank mRNA | AK028974.1 | - | 4 | 1664 | ||
| AK030704.1 | - | 4 | 1439 | |||
| AK048405.1 | - | 3 | 535 | |||
| AK053614.1 | - | 4 | 1674 | |||
| AK157918.1 | - | 4 | 1471 | |||
| AK172074.1 | - | 4 | 1672 | |||
| BC079846.1 | - | 4 | 1679 | 5707855 | ||
| BC113176.1 | - | 5 | 1579 | 40060773 | ||
| BC113781.1 | - | 3 | 642 | 40060770 | ||
| BC138457.1 | - | 4 | 1582 | 8861477 | ||
| BC138458.1 | - | 4 | 1582 | 8861478 | ||