Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc14a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171010.1 | NP_001164481.1 | 3 | 492 | ||
NM_001171011.1 | NP_001164482.1 | 2 | 227 | |||
NM_028122.4 | NP_082398.1 | 2 | 287 | |||
GenBank mRNA | AF448798.1 | - | 1 | 84 | ||
AJ420967.1 | - | 2 | 146 | |||
AK012066.1 | - | 2 | 137 | |||
AK041979.1 | - | 2 | 229 | |||
AK153891.1 | - | 3 | 397 | |||
BC058594.1 | - | 2 | 174 | 5364683 | ||
BC086673.1 | - | 2 | 183 | 30646772 | ||
BC100570.1 | - | 2 | 174 | 30044353 | ||
BY231473.1 | - | 2 | 287 |