Search
Search

Obfc1| Gene Symbol: Obfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175360.2 | NP_780569.1 | 5 | 701 | ||
| GenBank mRNA | AK075574.1 | - | 5 | 701 | ||
| AK135228.1 | - | 5 | 719 | |||
| AK146929.1 | - | 5 | 779 | |||
| AK164386.1 | - | 5 | 711 | |||
| BC029548.1 | - | 5 | 741 | 5012064 | ||