Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017366.3 | NP_059062.1 | 10 | 1056 | ||
GenBank mRNA | AF017176.1 | - | 7 | 704 | ||
AK076037.1 | - | 10 | 1017 | |||
AK146334.1 | - | 5 | 529 | |||
AK189988.1 | - | 1 | 30 | |||
AK208302.1 | - | 1 | 30 | |||
BC026559.1 | - | 10 | 1035 | 5037878 | ||
Z71189.1 | - | 10 | 977 |