Search
Search

Adk| Gene Symbol: Adk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243041.1 | NP_001229970.1 | 7 | 788 | ||
| NM_134079.4 | NP_598840.1 | 7 | 745 | |||
| GenBank mRNA | AK032871.1 | - | 7 | 703 | ||
| AK050135.1 | - | 1 | 48 | |||
| AK151863.1 | - | 7 | 790 | |||
| AK160264.1 | - | 7 | 742 | |||
| AY540996.1 | - | 7 | 687 | |||
| AY540997.1 | - | 6 | 632 | |||
| BC009659.1 | - | 7 | 710 | 3485959 | ||