Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007435.1 | NP_031461.1 | 9, 10 | 2439 | ||
GenBank mRNA | AK088792.1 | - | 9, 10 | 2309 | ||
BC011273.1 | - | 9, 10 | 2258 | 3710503 | ||
BC079840.1 | - | 9, 10 | 2317 | 6810147 | ||
Z33637.1 | - | 9, 10 | 2439 |