Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AXIN1 SIRNA Assays ›
          Gene Symbol:
          Axin1
          Gene Name:
          axin 1
          Gene Aliases:
          AI316800, Axin, Fu, Kb, Ki, fused, kinky, knobbly
          Chromosome Location:
          Chr.17: 26130972 - 26195811 on Build GRCm38
          UniGene ID:
          Mm.23684
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS202310
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Axin1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001159598.1 NP_001153070.1 2 730
          NM_009733.2 NP_033863.2 2 730
          GenBank mRNA AF009011.1 - 2 682
          BC011544.1 - 2 450
          BC113171.1 - 2 462 40061055

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_024405, AF017756.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          in utero embryonic development
          optic placode formation
          regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          nucleocytoplasmic transport
          apoptotic process
          signal transduction
          activation of JUN kinase activity
          multicellular organism development
          determination of left/right symmetry
          sensory perception of sound
          adult walking behavior
          cell death
          dorsal/ventral axis specification
          dorsal/ventral pattern formation
          Wnt signaling pathway
          hemoglobin metabolic process
          forebrain anterior/posterior pattern specification
          Wnt-activated signaling pathway involved in forebrain neuron fate commitment
          cell differentiation
          protein catabolic process
          negative regulation of Wnt signaling pathway
          positive regulation of transforming growth factor beta receptor signaling pathway
          olfactory placode formation
          cytoplasmic microtubule organization
          positive regulation of protein ubiquitination
          activation of protein kinase activity
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          erythrocyte homeostasis
          negative regulation of transcription elongation from RNA polymerase II promoter
          regulation of catenin import into nucleus
          post-anal tail morphogenesis
          termination of G-protein coupled receptor signaling pathway
          positive regulation of JUN kinase activity
          positive regulation of GTPase activity
          cellular protein complex assembly
          negative regulation of fat cell differentiation
          positive regulation of protein catabolic process
          positive regulation of protein kinase activity
          positive regulation of transcription, DNA-templated
          positive regulation of JNK cascade
          embryonic eye morphogenesis
          axial mesoderm development
          axial mesoderm formation
          neurological system process
          negative regulation of protein metabolic process
          protein homooligomerization
          positive regulation of ubiquitin-protein transferase activity
          muscle cell development
          canonical Wnt signaling pathway
          embryonic skeletal joint morphogenesis
          canonical Wnt signaling pathway involved in neural plate anterior/posterior pattern formation
          cellular response to organic cyclic compound
          genetic imprinting
          negative regulation of canonical Wnt signaling pathway
          Wnt signaling pathway involved in somitogenesis
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          p53 binding
          signal transducer activity
          GTPase activator activity
          receptor binding
          protein binding
          beta-catenin binding
          protein C-terminus binding
          enzyme binding
          protein kinase binding
          protein domain specific binding
          receptor signaling complex scaffold activity
          ubiquitin protein ligase binding
          protein complex scaffold
          identical protein binding
          protein homodimerization activity
          protein self-association
          SMAD binding
          armadillo repeat domain binding
          I-SMAD binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0