Search
Search

Ccne2| Gene Symbol: Ccne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037134.2 | NP_001032211.1 | 6 | 638 | ||
| NM_001282943.1 | NP_001269872.1 | 6 | 551 | |||
| NM_009830.3 | NP_033960.1 | 6 | 548 | |||
| GenBank mRNA | AF091432.1 | - | 5 | 443 | ||
| AF106691.1 | - | 4 | 418 | |||
| AK010929.1 | - | 6 | 478 | |||
| AK011883.1 | - | 1 | 110 | |||
| AK031116.1 | - | 6 | 475 | |||
| AK037791.1 | - | 6 | 478 | |||
| AK158720.1 | - | 6 | 1064 | |||
| AK167428.1 | - | 6 | 478 | |||
| BC053727.1 | - | 5 | 438 | 30061057 | ||