Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009900.2 | NP_034030.2 | 10 | 1682 | ||
GenBank mRNA | AF097415.1 | - | 10 | 1160 | ||
AF139724.1 | - | 10 | 1111 | |||
AK141656.1 | - | 11 | 2318 | |||
AK155104.1 | - | 10 | 976 | |||
AK165000.1 | - | 10 | 1210 | |||
AK171231.1 | - | 10 | 976 | |||
AK172148.1 | - | 7 | 1665 | |||
BC120699.1 | - | 10 | 1073 | 40129622 | ||
BC137625.1 | - | 10 | 1073 | 8860644 |