Search
Search

Cpt1a| Gene Symbol: Cpt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013495.2 | NP_038523.2 | 13 | 1586 | ||
| GenBank mRNA | AF017175.1 | - | 1474 | |||
| AK077454.1 | - | 13 | 1540 | |||
| AK128925.1 | - | 13 | 1540 | |||
| AK133472.1 | - | 13 | 1541 | |||
| AK136487.1 | - | 13 | 1540 | |||
| AK147770.1 | - | 13 | 1541 | |||
| AK149688.1 | - | 13 | 1537 | |||
| BC038395.1 | - | 12 | 1521 | 2649352 | ||
| BC046383.1 | - | 12 | 1521 | 4160676 | ||
| BC054791.1 | - | 13 | 1567 | 6832731 | ||
| D49472.1 | - | 12 | 1513 | |||