Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277219.1 | NP_001264148.1 | 3 | 791 | ||
NM_001277221.1 | NP_001264150.1 | 2 | 425 | |||
NM_133656.5 | NP_598417.2 | 3 | 961 | |||
GenBank mRNA | AK028179.1 | - | 3 | 768 | ||
AK028488.1 | - | 3 | 911 | |||
AK036905.1 | - | 3 | 742 | |||
AK052795.1 | - | 3 | 750 | |||
AK076097.1 | - | 3 | 656 | |||
AK088519.1 | - | 3 | 762 | |||
AK139089.1 | - | 3 | 747 | |||
AK152549.1 | - | 3 | 935 | |||
AK154987.1 | - | 3 | 946 | |||
AK161761.1 | - | 3 | 903 | |||
AK163291.1 | - | 2 | 400 | |||
AK198502.1 | - | 2 | 110 | |||
AK200050.1 | - | 2 | 86 | |||
AK202550.1 | - | 2 | 46 | |||
AK212188.1 | - | 2 | 97 | |||
BC012216.1 | - | 3 | 897 | 3992341 | ||
BC031149.1 | - | 3 | 724 | 4975654 | ||
S72408.1 | - | 3 | 886 |