Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddost | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007838.2 | NP_031864.2 | 6 | 765 | ||
GenBank mRNA | AK077651.1 | - | 6 | 627 | ||
AK081542.1 | - | 6 | 657 | |||
AK084267.1 | - | 6 | 649 | |||
AK148095.1 | - | 6 | 646 | |||
AK150683.1 | - | 6 | 656 | |||
AK200087.1 | - | 4 | 326 | |||
BC068132.1 | - | 6 | 765 | 6808198 | ||
D89063.1 | - | 6 | 767 |