Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010143.1 | NP_034273.1 | 8 | 2070 | ||
GenBank mRNA | AK144414.1 | - | 8 | 2086 | ||
AK158043.1 | - | 9 | 3010 | |||
BC014822.1 | - | 8 | 2071 | 3673003 | ||
BC027805.1 | - | 5 | 1253 | |||
BC053085.1 | - | 8 | 2070 | 5708827 | ||
U11493.1 | - | 6 | 1530 |