Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290414.1 | NP_001277343.1 | 7 | 1008 | ||
NM_001290415.1 | NP_001277344.1 | 5 | 519 | |||
NM_010200.3 | NP_034330.2 | 5 | 1003 | |||
GenBank mRNA | AF020737.1 | - | 5 | 636 | ||
AK141848.1 | - | 5 | 519 | |||
BC018238.1 | - | 5 | 1003 | 4481123 | ||
U66202.1 | - | 5 | 630 |