Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FYN SIRNA Assays ›
          Gene Symbol:
          Fyn
          Gene Name:
          Fyn proto-oncogene
          Gene Aliases:
          AI448320, AW552119
          Chromosome Location:
          Chr.10: 39369770 - 39565381 on Build GRCm38
          UniGene ID:
          Mm.4848
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS204459
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fyn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122892.1 NP_001116364.1 7 1017
          NM_001122893.1 NP_001116365.1 8 1087
          NM_008054.2 NP_032080.2 8 1087
          GenBank mRNA AK156584.1 - 8 1089
          AK171646.1 - 8 1089
          BC010554.1 - 7 700
          BC032149.1 - 7 732 3602546
          BC092217.1 - 7 1013 6403711
          BQ179147.1 - 4 286
          M27266.1 - 9 799
          U70324.1 - 8 800

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          immune system process
          protein phosphorylation
          cell surface receptor signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          multicellular organism development
          regulation of cell shape
          negative regulation of gene expression
          positive regulation of neuron projection development
          positive regulation of phosphatidylinositol 3-kinase signaling
          phosphorylation
          peptidyl-tyrosine phosphorylation
          cell differentiation
          forebrain development
          negative regulation of protein ubiquitination
          cellular response to platelet-derived growth factor stimulus
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          negative regulation of protein catabolic process
          myelination
          regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of neuron apoptotic process
          innate immune response
          response to ethanol
          protein autophosphorylation
          dendrite morphogenesis
          activated T cell proliferation
          T cell receptor signaling pathway
          detection of mechanical stimulus involved in sensory perception of pain
          cellular response to peptide hormone stimulus
          cellular response to transforming growth factor beta stimulus
          positive regulation of protein localization to nucleus
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          nucleotide binding
          G-protein coupled receptor binding
          glycoprotein binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          protein binding
          ATP binding
          tubulin binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein complex binding
          T cell receptor binding
          CD4 receptor binding
          CD8 receptor binding
          phosphatidylinositol 3-kinase binding
          ion channel binding
          metal ion binding
          ephrin receptor binding
          peptide hormone receptor binding
          growth factor receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0