Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GCLC SIRNA Assays ›
          Gene Symbol:
          Gclc
          Gene Name:
          glutamate-cysteine ligase, catalytic subunit
          Gene Aliases:
          D9Wsu168e, GLCL-H, Ggcs-hs, Glclc
          Chromosome Location:
          Chr.9: 77754516 - 77794489 on Build GRCm38
          UniGene ID:
          Mm.89888
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS204720
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gclc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010295.2 NP_034425.1 4, 5 836
          GenBank mRNA AK144339.1 - 4, 5 837
          AK160876.1 - 4, 5 596
          AK169676.1 - 4, 5 809
          BC019374.1 - 4, 5 593 4195425
          U85414.1 - 4, 5 561
          U85498.1 - 4, 5 561

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          cysteine metabolic process
          glutamate metabolic process
          glutathione metabolic process
          glutathione biosynthetic process
          response to oxidative stress
          apoptotic mitochondrial changes
          response to heat
          response to xenobiotic stimulus
          response to hormone
          L-ascorbic acid metabolic process
          negative regulation of protein ubiquitination
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          cell redox homeostasis
          negative regulation of transcription, DNA-templated
          response to arsenic-containing substance
          regulation of blood vessel size
          response to nitrosative stress
          regulation of mitochondrial depolarization
          negative regulation of extrinsic apoptotic signaling pathway
          nucleotide binding
          magnesium ion binding
          glutamate-cysteine ligase activity
          ATP binding
          glutamate binding
          ligase activity
          ADP binding
          protein heterodimerization activity
          coenzyme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0