Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142706.1 | NP_001136178.1 | 13 | 2115 | ||
NM_008198.2 | NP_032224.2 | 13 | 2115 | |||
GenBank mRNA | AK098094.1 | - | 13 | 1775 | ||
AK149531.1 | - | 13 | 1748 | |||
AK149573.1 | - | 13 | 1774 | |||
AK151220.1 | - | 7 | 734 | |||
AK151618.1 | - | 7 | 734 | |||
AK151974.1 | - | 7 | 734 | |||
BC005451.1 | - | 13 | 1749 | 3591669 | ||
K01496.1 | - | 8 | 855 | |||
K01498.1 | - | 3 | 279 | |||
M57890.1 | - | 13 | 1818 |