Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008310.3 | NP_032336.1 | 3 | 985 | ||
GenBank mRNA | AK045364.1 | - | 3 | 1054 | ||
BC120507.1 | - | 3 | 841 | 40129430 | ||
BC137604.1 | - | 3 | 840 | 8860623 | ||
Z14224.1 | - | 3 | 984 |
Set Membership: |
G-protein coupled receptor |