Search
Search

| Gene Symbol: Icam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008319.2 | NP_032345.2 | 9 | 2092 | ||
| GenBank mRNA | BC113140.1 | - | 9 | 2040 | 40048010 | |
| U06483.1 | - | 9 | 2040 | |||
| Gene Symbol: Gm26592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082527.1 | - | 883 | |||