Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008325.4 | NP_032351.2 | 14 | 2213 | ||
NM_172997.1 | NP_766585.1 | 13 | 2072 | |||
GenBank mRNA | AK031414.1 | - | 13 | 1867 | ||
AK045330.1 | - | 12 | 1700 | |||
AK155627.1 | - | 14 | 2214 | |||
AK169998.1 | - | 13 | 1871 | |||
AK171124.1 | - | 9 | 1223 | |||
BC057935.1 | - | 13 | 1834 | 5327150 | ||
L34111.1 | - | 13 | 1787 |