Search
Search

Il10ra| Gene Symbol: Il10ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008348.2 | NP_032374.1 | 2, 3 | 262 | ||
| GenBank mRNA | AK137663.1 | - | 2, 3 | 263 | ||
| AK149680.1 | - | 2, 3 | 257 | |||
| AK150022.1 | - | 2, 3 | 259 | |||
| AK150524.1 | - | 2, 3 | 266 | |||
| AK153120.1 | - | 2, 3 | 266 | |||
| AK159566.1 | - | 2, 3 | 259 | |||
| L12120.1 | - | 2, 3 | 267 | |||