Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080926.1 | NP_001074395.1 | 13 | 1845 | ||
GenBank mRNA | AJ312058.1 | - | 13 | 1691 | ||
AK030143.1 | - | 10 | 1355 | |||
AK038580.1 | - | 12 | 1882 | |||
BC127061.1 | - | 14 | 1598 | |||
D85463.1 | - | 14 | 2221 |