Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289729.1 | NP_001276658.1 | 6 | 847 | ||
NM_133213.2 | NP_573476.2 | 6 | 847 | |||
GenBank mRNA | AF367247.1 | - | 6 | 511 | ||
AF428102.1 | - | 6 | 698 | |||
AK078835.1 | - | 6 | 654 | |||
AK085520.1 | - | 6 | 645 | |||
AK087627.1 | - | 6 | 654 | |||
AK164538.1 | - | 6 | 675 | |||
AK172086.1 | - | 6 | 848 | |||
BC140977.1 | - | 6 | 491 | 9055589 |