Search
Search

Ly75| Gene Symbol: Ly75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013825.3 | NP_038853.2 | 26 | 3540 | ||
| GenBank mRNA | AF395445.1 | - | 26 | 3540 | ||
| AK083406.1 | - | 595 | ||||
| BC150734.1 | - | 3561 | 9087645 | |||
| U19271.1 | - | 3574 | ||||