Search
Search

Ly75| Gene Symbol: Ly75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013825.3 | NP_038853.2 | 6, 7 | 1060 | ||
| GenBank mRNA | AF395445.1 | - | 6, 7 | 1060 | ||
| AK049301.1 | - | 6, 7 | 1103 | |||
| AK157921.1 | - | 6, 7 | 1124 | |||
| BC150734.1 | - | 1081 | 9087645 | |||
| U19271.1 | - | 1094 | ||||