Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161775.1 | NP_001155247.1 | 23 | 3050 | ||
NM_013607.2 | NP_038635.2 | 23 | 3050 | |||
GenBank mRNA | AK137226.1 | - | 24 | 3027 | ||
AK137716.1 | - | 23 | 3005 | |||
AK173045.1 | - | 23 | 3014 | |||
BC026142.1 | - | 23 | 3035 | 3989495 | ||
D85923.1 | - | 23 | 3005 | |||
D85924.1 | - | 23 | 3005 |