Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010882.3 | NP_035012.2 | 1 | 324 | ||
GenBank mRNA | AK005078.1 | - | 1 | 282 | ||
AK075729.1 | - | 1 | 282 | |||
AK075735.1 | - | 1 | 270 | |||
AK076025.1 | - | 1 | 280 | |||
AK078129.1 | - | 1 | 280 | |||
AK135444.1 | - | 1 | 280 | |||
AK149086.1 | - | 1 | 280 | |||
AU080413.1 | - | 1 | 335 | |||
BC147249.1 | - | 1 | 305 | 9056296 | ||
BC147250.1 | - | 1 | 305 | 9056298 | ||
M80840.1 | - | 1 | 260 |